Merck
  • Human Striatal Dopaminergic and Regional Serotonergic Synaptic Degeneration with Lewy Body Disease and Inheritance of APOE ε4.
Human Striatal Dopaminergic and Regional Serotonergic Synaptic Degeneration with Lewy Body Disease and Inheritance of APOE ε4.
The American journal of pathology (2017-02-19)
Nadia Postupna, Caitlin S Latimer, Eric B Larson, Emily Sherfield, Julie Paladin, Carol A Shively, Matthew J Jorgensen, Rachel N Andrews, Jay R Kaplan, Paul K Crane, Kathleen S Montine, Suzanne Craft, C Dirk Keene, Thomas J Montine
ZUSAMMENFASSUNG
Cognitive impairment in older individuals is a complex trait that in population-based studies most commonly derives from an individually varying mixture of Alzheimer disease, Lewy body disease, and vascular brain injury. We investigated the molecular composition of synaptic particles from three sources: consecutive rapid autopsy brains from the Adult Changes in Thought Study, a population-based cohort; four aged nonhuman primate brains optimally processed for molecular investigation; and targeted replacement transgenic mice homozygous for APOE ε4. Our major goal was to characterize the molecular composition of human synaptic particles in regions of striatum and prefrontal cortex. We performed flow cytometry to measure six markers of synaptic subtypes, as well as amyloid β 42 and paired helical filament tau. Our results showed selective degeneration of dopaminergic terminals throughout the striatum in individuals with Lewy body disease, and serotonergic degeneration in human ventromedial caudate nucleus from individuals with an APOE ε4 allele. Similar results were seen in mouse caudate nucleus homozygous for APOE ε4 via targeted replacement. Together, extension of these clinical, pathologic, and genetic associations from tissue to the synaptic compartment of cerebral cortex and striatum strongly supports our approach for accurately observing the molecular composition of human synapses by flow cytometry.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-SLC6A4/5HTT/SERT antibody produced in goat, affinity isolated antibody, buffered aqueous solution