Direkt zum Inhalt
Merck
Anti-RPS13 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-RPS13 antibody produced in rabbit

Anti-RPS13 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(e):
40S ribosomal protein S13, Anti-Ribosomal protein S13, Anti-S13

Products

Produktkategorie
Marke
Biologische Quelle
Klonalität
Gen-Alias
Physikalische Form
Produktreihe
Antikörperform
Forschungsbereich
Konjugat
Posttranslationale Modifikation
Speziesreaktivität
Verfahren