Über diesen Artikel
NACRES:
NB.01
UNSPSC Code:
41121500
Volume:
0.1-3 μL
Packaging:
pack of 1 ea
Feature:
electronic: no, single-channel
Manufacturer/tradename:
Sartorius 728010
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenfeature
electronic: no, single-channel
packaging
pack of 1 ea
manufacturer/tradename
Sartorius 728010
volume
0.1-3 μL
General description
The Sartorius Proline Plus pipettes of the Biohitfamily are high quality with a light pipetting force. With robust design, the Proline Plus is suitable for variable every day laboratory applications and covers the full range of 0.1μL to 10 mL.
- Reduced pipetting forces prevent RSI and improve the results
- Accurate and precise
- Contamination free pipetting with Safe-Cone Filters
- Robust design
- Easiest pipette to maintain
Legal Information
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
Still not finding the right product?
Explore all of our products under Sartorius Proline® Plus mechanical pippettor
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?