Über diesen Artikel
NACRES:
NB.35
UNSPSC Code:
46181503
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltensize
L
manufacturer/tradename
Arco 1724205
General description
MICROGARD™ 2500 STANDARD is designed to protect from saturating liquid spray and particulates, the fabric′s physical strength and flexibility ensure you are protected yet comfortable.
Approved to:
EN14605 Type 4 Spray-tight
EN ISO 13982-1 Type 5 Particulate protection
EN13034 Type 6 Reduced spray
EN1073-2 Barrier to radioactive particulates - Class 2
EN14126 Barrier to infective agents
EN1149-1 Anti-static
Approved to:
EN14605 Type 4 Spray-tight
EN ISO 13982-1 Type 5 Particulate protection
EN13034 Type 6 Reduced spray
EN1073-2 Barrier to radioactive particulates - Class 2
EN14126 Barrier to infective agents
EN1149-1 Anti-static
Features and Benefits
- 3 piece hood
- Ultrasonically welded seamsFinger loops
- 2-way front zip with resealable storm flap
- Elasticated hood, waist, wrists and ankles
Legal Information
Microgard is a trademark of Microgard Limited
Still not finding the right product?
Explore all of our products under MICROGARD™ 2500 coveralls for protection from saturating liquid spray
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?