Über diesen Artikel
NACRES:
NB.35
UNSPSC Code:
46181503
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltensize
M (elasticated ankles)
manufacturer/tradename
Arco 1762004
General description
MICROGARD™ 2000 fully elasticated coverall with hood , setting the standard in Type 5/6 performance and protection, offers protection from infective agents, pesticides, particulates, fibres and liquid chemical spray.
Approved to:
EN ISO 13982-1 Type 5 Particulate protection
EN13034 Type 6 Reduced spray
EN1073-2 Barrier to radioactive particulates - Class 2
EN14126 Barrier to infective agents
EN1149-5 Anti-static
Approved to:
EN ISO 13982-1 Type 5 Particulate protection
EN13034 Type 6 Reduced spray
EN1073-2 Barrier to radioactive particulates - Class 2
EN14126 Barrier to infective agents
EN1149-5 Anti-static
Features and Benefits
- Features: 3-piece hood
- 2-way front zipper with re-sealable storm flap
- Finger loops
- Elasticated hood, waist, wrists and ankles
Legal Information
Microgard is a trademark of Microgard Limited
Still not finding the right product?
Explore all of our products under MICROGARD™ 2000 coveralls for protection from infective agent
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?