Merck
Z659622
Bandelin SONOREX Ultrasonic bath accessories
tank lid to fit Z659576, Z659002 and Z659592
Synonym(e):
Bandelin 3003, lid, protective lid, tank cover, ultrasonic cover
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Über diesen Artikel
NACRES:
NB.74
UNSPSC Code:
41103706
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
stainless steel
description
Bandelin D 100
packaging
pack of 1 ea
manufacturer/tradename
Bandelin 3003
availability
not available in USA and Canada
General description
Stainless steel lid for an ultrasonic bath protects the bath liquid from external contamination and prevents unpleasant odors from the cleaning or disinfection agents added. In addition, the noise emitted when the ultrasonic cleaner is in operation, which may be perceived as annoying, is reduced. The lids are designed in such a way that any condensation that occurs is directed back into the bath.
Features and Benefits
  • protection of the bath fluid from contamination
  • sound-absorbing
  • shortens the heating phase
Legal Information
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)