Merck
Z376779
Lazy-L Spreader
10 per pouch
Synonym(e):
L shape spreader, bacterial spreader, disposable cell spreader
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Über diesen Artikel
NACRES:
NB.22
UNSPSC Code:
41161502
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
polystyrene
sterility
sterile
packaging
case of 500 10 per pack
manufacturer/tradename
Excel Scientific SPR-L-S10, Lazy-L Spreader
General description
Lazy-L Spreader disposable microbiological spreaders provide an economical alternative to bending glass rods or pipets for spreading samples on agar surfaces and filters in Petri dishes. They are supplied sterile -- no need to flame sterilize or autoclave.
  • unlike glass spreaders, durable polystyrene Lazy-L Spreaders are non-breakable and pose no safety hazard
  • L-shaped for convenient "lazy" spreading; just turn the Petri dish for smooth, even sample distribution
  • smooth spreading surface and a slight upward turn reduce the chance that the spreader will dig into agar
  • presterilization eliminates flaming or autoclaving and reduces risk of contamination
  • packaged in presterilized pouches of one or ten each
  • no more time spent bending glass rods or waiting for glass or metal spreaders to cool before use!
Legal Information
Lazy-L Spreader is a trademark of Excel Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)