Ansicht ändern
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
10 ea | Warenkorb auf Verfügbarkeit prüfen | € 36,00 |
Über diesen Artikel
UNSPSC Code:
41103806
NACRES:
NB.72
eCl@ss:
32100100
Material:
PTFE-coated , cylindrical
Manufacturer/tradename:
BRAND 137113
€ 36,00
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenQuality Segment
material
PTFE-coated , cylindrical
manufacturer/tradename
BRAND 137113
L
10 mm
bar diam.
6 mm
General description
BRAND® magnetic cylindrical stirring bars are expertly designed for optimal mixing performance, featuring a smooth, round shape that enhances stirring efficiency. Their Alnico V magnetic cores are protected by a robust PTFE (polytetrafluoroethylene) coating, ensuring durability and resistance to chemical damage. These stirring bars are particularly effective for use in large volumes or thick mixtures, making them ideal for a variety of laboratory applications.
Features and Benefits
- BRAND® magnetic cylindrical stirring bars are round with a smooth surface.
- The Alnico V magnetic cores are encapsulated with high-grade, crack-resistant PTFE (polytetrafluoroethylene) coating.
- Ideal for large volumes or highly viscous mixtures.
Other Notes
BR137113 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| manufacturer/tradename BRAND 137113 | manufacturer/tradename BRAND 137114 | manufacturer/tradename BRAND 137109 | manufacturer/tradename BRAND 137111 |
| material PTFE-coated , cylindrical | material PTFE-coated , cylindrical | material PTFE-coated , cylindrical | material PTFE-coated , cylindrical |
| L 10 mm | L 15 mm | L 13 mm | L 15 mm |
| bar diam. 6 mm | bar diam. 6 mm | bar diam. 3 mm | bar diam. 1.7 mm |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
Still not finding the right product?
Explore all of our products under BRAND® Magnetrührstäbchen, PTFE, zylindrisch
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?