Direkt zum Inhalt
Z326461
BRAND® BLAUBRAND® Messkolben, PP-Stopfen, Braunglas
volume 250 mL, accuracy: 0.15 mL
Synonym(e):
amber flask, analytical flask, calibration flask with stopper, measuring flask, volumetric flask
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Über diesen Artikel
NACRES:
NB.43
UNSPSC Code:
41121804
Capacity:
250 mL
Material:
borosilicate glass 3.3 flask
Packaging:
pack of 2 ea
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
borosilicate glass 3.3 flask
joint
joint ST/NS neck14/23
description
calibrated to contain (TC, IN)
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
packaging
pack of 2 ea
manufacturer/tradename
BRAND 37409
volume measuring range
250 mL, accuracy: 0.15 mL
capacity
250 mL
General description
The BLAUBRAND® volumetric flask combines precision and reliability, featuring a square grip PP stopper for easy handling. Crafted from high-quality Boro 3.3 glass and calibrated to contain (TC, In), each flask is marked with high-contrast white enamel for clear readability. Supplied with a batch number and certificate, these flasks ensure quality and traceability, making them ideal for laboratory use.
Other Notes
BR37409 (India)
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)