Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
100 ea | Warenkorb auf Verfügbarkeit prüfen | € 840,00 |
Über diesen Artikel
€ 840,00
material
polystyrene
packaging
case of 100 ea
manufacturer/tradename
Nunc 167064
L × W
128 mm × 85 mm
wells
8
General description
Application
Features and Benefits
- Flat well bottom and stackable.
- Raised well rims lower risk of cross contamination.
- Sterilized by gamma irradiation and certified to be non-pyrogenic.
- Excellent optical quality.
- Alphanumerical codes for well identification.
- Vented lids for gas exchange.
- Recessed areas around wells act as a water reservoir to reduce evaporation from wells.
- Lids with notched corners for easy orientation.
- Large culture area.
- Accept standard glass cover slips: 24 x 60 mm for 4-well plate and 22 x 22 mm for 8-well plate.
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description wells 8, polystyrene, L × W 128 mm × 85 mm, witht lid, Nunclon® treated, case of 100 ea | description wells 6, polystyrene, L × W 128 mm × 86 mm, with lid, non-treated, case of 75 ea | description polystyrene, wells 6, without lid, Nunclon® Delta treated, L × W 128 mm × 85 mm, case of 85 ea | description wells 24, polystyrene, L × W 128 mm × 86 mm, with lid, non-treated, case of 75 ea |
| manufacturer/tradename Nunc 167064 | manufacturer/tradename Nunc 150239 | manufacturer/tradename Nunc 140685 | manufacturer/tradename Nunc 144530 |
| material polystyrene | material polystyrene | material polystyrene | material polystyrene |
| L × W 128 mm × 85 mm | L × W 128 mm × 86 mm | L × W 128 mm × 85 mm | L × W 128 mm × 86 mm |
| packaging case of 100 ea | packaging case of 75 ea | packaging case of 85 ea | packaging case of 75 ea |
| wells 8 | wells 6 | wells 6 | wells 24 |
Still not finding the right product?
Explore all of our products under Nunc® Cell-Culture Treated Multidishes
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?