Merck
SMCXPRO
SMCxPRO® Immunoassay Instrument
High Sensitivity Protein Measurement at the fg/mL level
Synonym(e):
SMC, Singulex, single molecule counting, xPRO
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
UNSPSC Code:
41115401
SMCxPRO™ Immunoassay Instrument
The SMCxPRO™ system is the 2nd generation instrument supporting SMC™ technology assays. The SMCxPRO™ technology approach collects signal from analyte fluorochromes free-floating in suspension using a rotating laser, CCD camera optics, and individual photons are counted by Avalanche Photodiode (APD).
  • High sensitivity (femtogram/mL) quantification
  • Minimal bench space: 14” W x 16” H x 17.5” D
  • Affordable
  • Bead-based kits
SMC™ Immunoassay Kits
SMC™ Immunoassay Kits using Single Molecule Counting (SMC™) technology enable you to reach the highest levels of sensitivity possible in an immunoassay format.
  • SMC™ kits contain all reagents required to perform assays on the SMCxPRO™ Immunoassay System.
  • Configure with a proprietary paramagnetic microparticle solid phase capture for ultrasensitive biomarker quantification.
  • Standard immunoassay plate format is also available.
SMCxPRO and SMC are registered trademarks of Merck KGaA, Darmstadt, Germany
Melden Sie sich an, um fortzufahren.
Um weiterzulesen, melden Sie sich bitte an oder erstellen ein Konto.
Sie haben kein Konto?