Über diesen Artikel
UNSPSC Code:
41100000
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
polystyrene
description
flat bottom
storage temp.
2-8°C
General description
SigmaScreen Streptavidin coated high capacity plates utilize a proprietary coating technology which provides substantially greater biotin binding capacity than standard Streptavidin or ExtrAvidin coated plates. This clear, 96-well plate has a format of breakable 8-well strips on a single-well holding frame, and has a binding capacity of ≥ 15 pmol/well. Streptavidin coated high capacity plates provide a platform which allows the captured protein to be eluted for post-capture analysis by various methods such as MALDI, ICAT or SDS-PAGE.
Legal Information
SigmaScreen is a trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?