Über diesen Artikel
NACRES:
NB.35
UNSPSC Code:
41122804
Manufacturer/tradename:
Diversified Biotech NORC-1000
Packaging:
pack of 1 ea
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenpackaging
pack of 1 ea
manufacturer/tradename
Diversified Biotech NORC-1000
color
red
General description
NovaRack provides universal housing for test tubes and vials of different sizes on the same surface. The numeric labeling for each tube cavity allows one to easily organize and track up to 46 tubes. It accommodates 24 microcentrifuge tubes (0.5-2 mL), 14 test tubes (12 x 75 mm culture tubes, cryovials and a variety of blood collection tubes), 7 centrifuge tubes (15 mL) and one 50 mL tube. NovaRack is constructed of polypropylene, is autoclavable and is available in five colors. With the addition of the Carousel Extension rod, the space saving ability is doubled and the top rack is allowed to spin.
Legal Information
Novarack is a trademark of Diversified Biotech
Still not finding the right product?
Explore all of our products under NovaRack™
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?