Über diesen Artikel
NACRES:
NB.22
UNSPSC Code:
41121702
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
natural , round bottom
sterility
non-sterile
manufacturer/tradename
Thermo Scientific AB-0700
technique(s)
PCR: suitable
capacity
0.2 mL
General description
Precision moulded from virgin polypropylene and clean room produced.
Certified free from DNase, RNase and human genomic DNA contamination.
Certified free from DNase, RNase and human genomic DNA contamination.
Skirted low-profile plate
Suitable for 0.2 mL thermal cycler blocks
0.2 mL maximum well volume (when used with adhesive and heat seals)
Raised well rims for effective sealing and reduced cross-contamination risk
Sealable using mats, films and foils or cap strips
Suitable for 0.2 mL thermal cycler blocks
0.2 mL maximum well volume (when used with adhesive and heat seals)
Raised well rims for effective sealing and reduced cross-contamination risk
Sealable using mats, films and foils or cap strips
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?