Merck
OH30456415
Ohaus® Navigator® Portable Balance
Model NV2201, weighing capacity 2200 g, resolution: 0.1 g, AC/DC input 110 V, US 3-pin plug (North America)
Synonym(e):
Ohaus 30456415, digital scale, electronic scale device, laboratory balance, laboratory scales, scale, weighing balance, weighing machine, weighing scales, weight measuring device
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Über diesen Artikel
UNSPSC Code:
41111502
Feature:
CSA approved
Material:
metal weighing plate
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
metal weighing plate
feature
CSA approved
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30456415
weighing capacity measuring range
2200 g, resolution: 0.1 g
weighing pan W × L
14.4 cm × 19 cm
General description
OHAUS raises the bar in price-performance ratio with the newly-designed Navigator series which is perfect for general weighing applications in laboratory, industrial and education settings. Ruggedly constructed and multifunctional, the Navigator′s a powerful balance that can handle a diverse range of weighing applications with features such as fast stabilization time, overload protection at four times its rated capacity, and easy operation.
Application
Weighing, Parts Counting, Percent Weighing, Checkweighing
Features and Benefits
Menu and calibration lock switch, mechanical and software overload/underload protection, stability indicator, low battery indicator, auto shut-off, user-selectable print options, user-selectable communication settings
Legal Information
Navigator is a registered trademark of Ohaus Corp.
Ohaus is a registered trademark of Ohaus Corp.
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)