Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
1000 ea | Warenkorb auf Verfügbarkeit prüfen | € 85,60 |
Über diesen Artikel
€ 85,60
material
blue , polypropylene
sterility
sterile
packaging
pack of 1000 ea
manufacturer/tradename
Heathrow Scientific HS81121A
loop volume
1 μL
General description
Features and Benefits
- Use loop end to uniformly streak a gel surface
- Use needle end for removal of colonies
- Constructed of flexible polypropylene
- Sterile, 20 per bag with lot-to-lot traceability
- Disposable
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description loop volume 1 μL, flexible , sterile, blue | description loop volume 1 μL, Rigid, sterile, blue | description loop volume 10 μL, flexible , sterile, blue | description loop volume 10 μL, Rigid, sterile, blue |
| material blue , polypropylene | material acrylonitrile-butadiene-styrene (ABS) , blue | material blue , polypropylene | material acrylonitrile-butadiene-styrene (ABS) , blue |
| manufacturer/tradename Heathrow Scientific HS81121A | manufacturer/tradename Heathrow Scientific HS81121B | manufacturer/tradename Heathrow Scientific HS81121C | manufacturer/tradename Heathrow Scientific HS81121D |
| loop volume 1 μL | loop volume 1 μL | loop volume 10 μL | loop volume 10 μL |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| packaging pack of 1000 ea | packaging pack of 1000 ea | packaging pack of 1000 ea | packaging pack of 1000 ea |
Still not finding the right product?
Explore all of our products under Inoculating Needle/Loops
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?