Ansicht ändern
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
100 ea | Warenkorb auf Verfügbarkeit prüfen | € 23,90 |
Über diesen Artikel
NACRES:
NB.71
UNSPSC Code:
41122403
€ 23,90
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
polystyrene , white
size
large
sterility
non-sterile
packaging
pack of 100 ea
manufacturer/tradename
Heathrow Scientific 120656
L × W × H
127 mm × 88.9 mm × 0.5 mm
General description
The dual purpose scoop is a unique economical solution that combines the function of a scoop and a weigh boat into oneproduct. The weighing process has been streamlined, now just sample or retrieve the material in the scoop and place it directly on the scale. This eliminates the need for a weigh boat or weighing paper and saves valuable space in the lab. When finished the dual solution scoop is disposable.
Features and Benefits
- Made from anti-static polystyrene that is waterproof andreduces the chance of materials sticking to the scoop
- Ships and stores flat and when ready to use, is easy to assemble by inserting the locking tab into the hole
- Locking tab feature enables a scoopshape to help retain materials
- Coated to stand up to powders, gels, and semi-solids
- Broad tip design keeps the scoop stable while on the scale and enables materialsto be easily scooped and dispensed
- Disposable
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| material polystyrene , white | material polystyrene , white | material black , polystyrene | material stainless steel |
| manufacturer/tradename Heathrow Scientific 120656 | manufacturer/tradename Heathrow Scientific 120657 | manufacturer/tradename Heathrow Scientific 120679 | manufacturer/tradename Bel-Art F36738-1700 |
| packaging pack of 100 ea | packaging pack of 100 ea | packaging pack of 100 ea | packaging pack of 1 ea |
| sterility non-sterile | sterility non-sterile | sterility sterile; electron beam irradiated | sterility - |
| size large | size small | size large | size - |
| L × W × H 127 mm × 88.9 mm × 0.5 mm | L × W × H 127 mm × 63.5 mm × 0.5 mm | L × W × H 127 mm × 88.9 mm × 0.5 mm | L × W × H - |
Still not finding the right product?
Explore all of our products under Dual Purpose Scoops
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?