Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 90,30 |
Über diesen Artikel
€ 90,30
feature
to hold 12 x 1.5/2 mL tubes
packaging
pkg of 1 ea
manufacturer/tradename
Heathrow Scientific 120622
parameter
12,500 rpm speed (max.), 9,783 × g max. RCF
compatibility
Heathrow Scientific HS10050
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description to hold, 12 x 1.5/2 mL tubes | description to hold, 6 x 5 mL tubes | description to hold, 0.2 mL tubes (4 x 8 Strip) | description to hold, 12 x 1.5/2 mL tubes |
| manufacturer/tradename Heathrow Scientific 120622 | manufacturer/tradename Heathrow Scientific 120623 | manufacturer/tradename Heathrow Scientific HS100504 | manufacturer/tradename Heathrow Scientific HS100503 |
| compatibility Heathrow Scientific HS10050 | compatibility Heathrow Scientific HS10050 | compatibility Heathrow Scientific HS10050 | compatibility - |
| feature to hold 12 x 1.5/2 mL tubes | feature to hold 6 x 5 mL tubes | feature to hold 0.2 mL tubes (4 x 8 Strip) | feature to hold 12 x 1.5/2 mL tubes |
| parameter 12,500 rpm speed (max.), 9,783 × g max. RCF | parameter 12,500 rpm speed (max.), 9,783 × g max. RCF | parameter - | parameter 9,800 × g max. RCF |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under MagFuge® High Speed Centrifuge Rotor
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?