Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
PE-LD , low-density polyethylene bottle, polypropylene cap, wide-mouth bottle, natural cap
joint
joint threaded
feature
label Ethanol, vented, with cap
packaging
pack of 6 ea
manufacturer/tradename
Heathrow Scientific HS120253
capacity
500 mL
General description
- Chemical Abstract Service (CAS) No.
- National Fire Protection Association (NFPA) codes and symbols
- Protective Equipment Symbol (PES)
- Target Organ Information and Hazard Codes.
Application
Features and Benefits
- Wide bottle mouth makes filling easier
- Non-metallic vent that prevents the risk of solvent leakage
- Closure and stem are molded in one piece to make them leak-proof
- Bottles are made from LDPE, with a PP closure with the exception of the bottle designed for Sodium Hypochlorite, which is a white LDPE bottle for light protection
- Six named chemicals, a self labeling and assorted pack of bottles to choose from
- 500 mL bottle size
- Supplied in packs of 6
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| capacity 500 mL | capacity 500 mL | capacity 500 mL | capacity 500 mL |
| manufacturer/tradename Heathrow Scientific HS120253 | manufacturer/tradename Heathrow Scientific HS120256 | manufacturer/tradename Heathrow Scientific HS120252 | manufacturer/tradename Heathrow Scientific HS120254 |
| feature label Ethanol, with cap, vented | feature label blank label, vented, with cap | feature label Isopropanol, vented, with cap | feature label Methanol, vented, with cap |
| material PE-LD , natural cap, wide-mouth bottle, low-density polyethylene bottle, polypropylene cap | material PE-LD , low-density polyethylene bottle, natural cap, polypropylene cap, wide-mouth bottle | material PE-LD , low-density polyethylene bottle, polypropylene cap, wide-mouth bottle, yellow cap | material PE-LD , green cap, low-density polyethylene bottle, polypropylene cap, wide-mouth bottle |
| packaging pack of 6 ea | packaging pack of 6 ea | packaging pack of 6 ea | packaging pack of 6 ea |
| joint joint threaded | joint joint threaded | joint joint threaded | joint joint threaded |
Still not finding the right product?
Explore all of our products under Wide mouth safety labelled wash bottle
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?