Merck
GE29-0187-51
Unicorn® start 1.0 software, for use with ÄKTA® start
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Über diesen Artikel
UNSPSC Code:
41121800
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
manufacturer/tradename
Cytiva 29-0187-51
availability
available only in Europe
General description
  • Unicorn start 1.0 is the control software designed for the ÄKTA start
  • Intuitive design: simple and flexible method creation using predefined templates and phases (steps)
  • Interactive process picture: enables real-time monitoring of the run and an intuitive way to control the run
  • Easy reporting capabilities: evaluate, compare and share results, create and print pdf reports
  • Data Management: robust database storage with easy access to data, data security and data integrity
The Unicorn start software includes for different modules: System Control, Method Editor, Evaluation and Administration, allowing you to design runs, operate the ÄKTA start instrument and to avaulate and share results.
While ÄKTA start can function as a stand-alone system, Unicorn start software maximizes the performance of the instrument.
Legal Information
Unicorn is a registered trademark of Cytiva
ÄKTA is a registered trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)