Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
50 ea | Warenkorb auf Verfügbarkeit prüfen | € 1.100,00 |
Über diesen Artikel
€ 1.100,00
material
polystyrene plate, square wells, white
sterility
sterile
feature
lid: no
packaging
case of 50 ea
manufacturer/tradename
Corning 7249
size
1536 wells
well volume
12.5 μL
well working volume
1-8 μL
color
white
suitability
suitable for (cell cuture)
Features and Benefits
- Superior performance compared to other available commercial microplates: lower CV values, excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
- Optimized for automation with bar coding and standard readers
- Surface treated for optimal cell attachment
- Flat bottom wells with lid are compatible with automated dispensers including the PerkinElmer® FlexDrop
- Sterile and certified nonpyrogenic
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description white, high base, w/o lid, sterile | description black, clear bottom, low base, w/o lid, sterile | description black, high base, w/o lid, non-sterile | description black, with lid, sterile |
| suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) |
| size 1536 wells | size 1536 wells | size 1536 wells | size 1536 wells |
| material polystyrene plate, white , square wells | material clear wells, polystyrene plate, square wells | material black , polystyrene plate, square wells | material black , black bottom, polystyrene plate, square wells |
| feature lid: no | feature lid: no, black, clear bottom | feature lid: no | feature lid, skirt |
| well working volume 1-8 μL | well working volume 5-8 μL | well working volume 1-8 μL | well working volume 1-8 μL |
| manufacturer/tradename Corning 7249 | manufacturer/tradename Corning 3836 | manufacturer/tradename Corning 7246 | manufacturer/tradename Corning 7248 |
Still not finding the right product?
Explore all of our products under Corning® TC treated 1536 well microplates
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?