Fortfahren mit
CLS4099
Corning® Stripettor Ultra Pipet Controller
AC/DC input 120 - 240 V AC, universal plug set (Europlug, UK, US, and Australian Plugs)
Synonym(e):
Pipette Controller, Pipetting Device
zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Ihnen/SKUVerfügbarkeitPreis
Über diesen Artikel
NACRES:
NB.01
UNSPSC Code:
41121500
Manufacturer/tradename:
Corning 4099
Volume:
0.5-100 mL
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
manufacturer/tradename
Corning 4099
volume
0.5-100 mL
General description
This light, ergonomic, motorized pipet controller is designed for use with glass and plastic serological pipets from 0.5 to 100 mL. Conveniently positioned switches change operating modes and speeds quickly to handle different liquid volumes and various viscosities. Aspirating and dispensing speed is controlled through the finger triggers. When fully charged, the batteries enable up to 8 hours of continuous use. The large LCD display clearly indicates battery status, pipetting mode, and speed. Corning Stripettor Ultra Pipet Controller comes with a universal power supply, three 0.2 um filters, 3 colored nose pieces, a wall hanger and a charging stand.
Features and Benefits
  • LCD display for continuous visualization of speed, modes, and battery status
  • Colored nose pieces for user or application identification
Well positioned function buttons
  • Accessories: Universal charging adapter, 0.2um filter, nose -piece, wall-mounted holder
Legal Information
Corning is a registered trademark of Corning, Inc.
Stripettor is a trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)