Fortfahren mit
CLS4079
Corning® universal linear rack for Lambda® Plus single-and multi-channel pipettors
holds six single-channel pipettors, maximum of four 8-channel or 12-channel pipettors.
Synonym(e):
pipettor accessory
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
NACRES:
NB.01
UNSPSC Code:
41122801
Manufacturer/tradename:
Corning 4079
Feature:
Holds 4 x 8/12-channel pipetors, Holds 6 x single channel pipetors
Packaging:
pack of 1 ea
feature
Holds 4 x 8/12-channel pipetors, Holds 6 x single channel pipetors
packaging
pack of 1 ea
manufacturer/tradename
Corning 4079
General description
This universal stand is designed to hold six single-channel pipettors, maximum of four 8-channel or 12-channel pipettors.
Legal Information
Corning is a registered trademark of Corning, Inc.
Lambda is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)