Fortfahren mit
CLS3859BC
Corning® 1536 well plate, with generic barcode
black polystyrene plate, sterile, lid
Synonym(e):
1536 multiwell plate
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Ihnen/SKUVerfügbarkeitPreis
50 ea
Warenkorb auf Verfügbarkeit prüfen
€ 4.590,00
Über diesen Artikel
NACRES:
NB.13
UNSPSC Code:
41121800
Material:
black polystyrene plate, polystyrene plate
Size:
1536 wells
€ 4.590,00
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
black polystyrene plate, polystyrene plate
sterility
sterile
feature
lid
packaging
pack of 10, case of 50
maximum volume
12.5 μL
size
1536 wells
working volume
8 μL
suitability
suitable for (cell cuture)
General description
Bar Code Details
Corning barcode labels have been quality tested for optimal readability, chemical resistance and temperature variation.
  • Resistant to commonly used laboratory reagents (100% DMSO, Methanol, Ethanol, Ethyl Acetate, Methylene Chloride, Isopropanol, Acetronitrile, Ethanol/Water/TFA, Sodium Hydroxide, Hydrochloric Acid)
  • Resistant to temperature ranges of -80ºC to +52ºC
Technical Details
  • Bar Code labeling specifications
  • 2.75" x 0.25" label size
  • Code 128
  • Code 3 of 9
  • 10 mil narrow bar element (x-dimension = 0.010")
Generic barcoded Corning 1536 well microplates are now available. Code 128 bar code format ensures compatibility with most bar code scanning and software systems. To avoid mix-ups, the surface treatment of the microplate wells is easily identified in the human readable portion of the bar code.
  • Resistant to temperature ranges of -80ºC to +52ºC
  • 2.8 mm distance between well bottoms to plate bottom
  • Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
Microplate Features
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Superior flatness -minimizes variability within a plate and between plates
  • Flat bottom wells are compatible with automated dispensers
  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions.
  • Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface (binding capacity of approximately 100 to 200ng IgG/cm²)
  • Working volume of up to 10 μL
Bar Code Details
Corning barcode labels have been quality tested for optimal readability, chemical resistance and temperature variation.
  • Resistant to commonly used laboratory reagents (100% DMSO, Methanol, Ethanol, Ethyl Acetate, Methylene Chloride, Isopropanol, Acetronitrile, Ethanol/Water/TFA, Sodium Hydroxide, Hydrochloric Acid)
  • Resistant to temperature ranges of -80ºC to +52ºC
Technical Details
  • Bar Code labeling specifications
  • 2.75" x 0.25" label size
  • Code 128
  • Code 3 of 9
  • 10 mil narrow bar element (x-dimension = 0.010")
Legal Information
Corning is a registered trademark of Corning, Inc.
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
CLS3727BCCLS3729BCCLS3893BC
description
black polystyrene plate, sterile, lid
description
size 1536 wells, flat bottom white polystyrene wells, sterile
description
size 1536 wells, NBS surface, flat bottom white polystyrene wells
description
size 1536 wells, flat bottom clear polystyrene wells, black plate
size
1536 wells
size
1536 wells
size
1536 wells
size
1536 wells
suitability
suitable for (cell cuture)
suitability
suitable for (cell cuture)
suitability
suitable for (cell cuture)
suitability
suitable for (cell cuture)
material
polystyrene plate, black polystyrene plate
material
polystyrene plate, flat bottom white polystyrene wells
material
polystyrene plate, flat bottom white polystyrene wells
material
flat bottom clear polystyrene wells, polystyrene plate
working volume
8 μL
working volume
8 μL
working volume
8 μL
working volume
8 μL
maximum volume
12.5 μL
maximum volume
12.5 μL
maximum volume
12.5 μL
maximum volume
12.5 μL
feature
lid
feature
lid, skirt
feature
NBS surface, lid: no, skirt
feature
lid
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)