Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
2 ea | Warenkorb auf Verfügbarkeit prüfen | € 493,00 |
Über diesen Artikel
€ 493,00
material
polycarbonate (medical grade)
description
1/4 in. I.D. C-Flex® tubing with quick-connect
sterility
sterile
packaging
pack of 1, case of 2
manufacturer/tradename
Corning 3528
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| manufacturer/tradename Corning 3528 | manufacturer/tradename Corning 3558 | manufacturer/tradename Corning 3545 | manufacturer/tradename Corning 3569 |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile; γ-irradiated |
| material polycarbonate (medical grade) | material polycarbonate (medical grade) | material polycarbonate (medical grade) | material high-density polyethylene cap, polystyrene , screw top cap |
| packaging pack of 1, case of 2 | packaging pack of 1, case of 2 | packaging pack of 1, pack of 2 | packaging case of 6 |
| description 1/4 in. I.D. C-Flex® tubing with quick-connect | description 1/4 in. I.D. C-Flex® tubing with quick-connect | description 1/4 in. I.D. C-Flex® tubing with quick-connect | description - |
Still not finding the right product?
Explore all of our products under Corning® disposable spinner flasks, sidearm assembly
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?