Merck
AI Assist – BETA
Beschleunigen Sie Ihre Antworten mit Hilfe von KI
MMSANKITCVR
Cover for Milliflex Oasis® VHP resistant pump, internal flow sanitization kit
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Über diesen Artikel
NACRES:
NB.75
UNSPSC Code:
41122409
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
stainless steel
sterilization compatibility
autoclavable compatible
packaging
pkg of 1 unit
manufacturer/tradename
Milliflex®
technique(s)
Rapid Sterility Testing: suitable
application(s)
bioburden testing
pharmaceutical
sterility testing
compatibility
for use with Milliflex Oasis® sanitization kit for Milliflex Oasis VHP resistant pump
AI Assist – BETA
General description
Milliflex Oasis® system offers a solution to simplify and secure microbial testing process with premium levels of productivity, result reliability and regulatory compliance. The Milliflex Oasis® VHP resistant vacuum pump is a new generation of vacuum pump for filtration in VHP isolator environments. The Milliflex Oasis® VHP resistant pump, internal flow sanitization kit including its cover is an accessory to be used to decontaminate the interior of the Milliflex Oasis® VHP resistant pump.
Application
The Milliflex Oasis® system finds its application in bioburden testing and water monitoring of Pharmaceutical Products Analysis
Legal Information
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)