Direkt zum Inhalt
Merck

MABF2185

Anti-Dengue Virus 2 NS1 Antibody, clone 243

Synonym(e):

DENV NS1 protein

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern

Über diesen Artikel

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
monoclonal
Application:
Citations:
-
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


biological source

mouse

conjugate

unconjugated

antibody form

purified antibody

clone

monoclonal

mol wt

~48 kDa (calculated for NS1 protein; 378.71 kDa for the entire genome polyprotein.)

purified by

using protein G

species reactivity

virus

isotype

IgG1κ

UniProt accession no.

target post-translational modification

unmodified

General description

Dengue virus (DENV) is a mosquito-borne flavivirus and is one of the most common mosquito-transmitted viral infections. The DENV particle is about 500 nm in diameter and includes a positive-sense RNA genome with ~10,700 nucleotides and 3 structural proteins known as capsid (C), precursor membrane (prM), and envelope (E) proteins. In addition, the RNA genome of dengue virus also encodes seven nonstructural proteins (NS1, NS2A, NS2B, NS3, NS4A, NS4B and NS5) that are shown to be essential for viral replication. DENV NS1, a 48-kDa glycoprotein (aa 776-1127 of genome polyprotein), is highly conserved among all flaviviruses and is essential for viral replication. It is expressed as a monomer in infected cells and after post-translational modification in the lumen of endoplasmic reticulum, it forms homodimers associated with organelle membranes and the cell membrane. NS1 is reported to be involved in immune evasion, pathogenesis, and viral replication. Once cleaved off the polyprotein, is targeted to three destinations: the viral replication cycle, the plasma membrane and the extracellular compartment. NS1 is secreted in a hexamer form, which is composed of three dimers with a detergent-sensitive hydrophobic central cavity and it plays a role against host immune response. NS1 is shown to induce expression of sialidases, heparanase, and activates cathepsin L, which activates heparinase via enzymatic cleavage. These effects are may contribute to the endothelial hyperpermeability commonly observed in severe dengue disease. During the recovery phase, NS1 is cleared from the circulation by antibody-mediated effects.

Application

ELISA Analysis: A representative lot of this antibody detected Dengue Virus 2 NS1 in ELISA applications

Flow Cytometry Analysis: A representative lot of this antibody detected Dengue Virus 2 NS1 in Flow Cytometry applications

Analysis Note

Evaluated by ELISA with DENV2 NS1 recombinant protein.

Other Notes

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.




Analysenzertifikate (COA)

Suchen Sie nach Analysenzertifikate (COA), indem Sie die Lot-/Chargennummer des Produkts eingeben. Lot- und Chargennummern sind auf dem Produktetikett hinter den Wörtern ‘Lot’ oder ‘Batch’ (Lot oder Charge) zu finden.

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen