Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Über diesen Artikel
Empirische Formel (Hill-System):
C22H21ClN2O5
CAS-Nummer:
Molekulargewicht:
428.87
UNSPSC Code:
12352200
MDL number:
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenQuality Segment
assay
≥95% (NMR)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, protect from light
color
orange
solubility
DMSO: 50 mg/mL
shipped in
ambient
storage temp.
2-8°C
General description
A cell-permeable phenyl-pyraz-olylidene compound that competes with p53-derived peptide (aa 15-29) binding to MDMX in cell-free binding assays (EC50 = 2.3 µM) in a reversible manner and effectively disrupts cellular p53-MDMX interaction (IC50<20 µM in C33A, HER, and Weri1 cultures), while exhibiting much weaker affinity toward MDM2 (EC50 = 26 µM in p53 peptide competition assays). MDMX inhibition by 20 µM SJ-172550 and MDM2 inhibition by 5 µM Nutlin-3a (Cat. Nos. 444143 & 444151) are both shown to result in reduced proliferation and increased cellular caspase-3 in cultured cells, however, only MDM2, but not MDMX, inhibition results in a significant p53 protein level upregulation.
A cell-permeable phenyl-pyrazolylidene compound that competes with p53-derived peptide (aa 15-29) binding to MDMX in cell-free binding assays (EC50 = 2.3 M) in a reversible manner and effectively disrupts cellular p53-MDMX interaction (IC50<20 M in C33A, HER, and Weri1 cultures), while exhibiting much weaker affinity toward MDM2 (EC50 = 26 M in p53 peptide competition assays). MDMX inhibition by 20 M SJ-172550 and MDM2 inhibition by 5 M Nutlin-3a are both shown to result in reduced proliferation and increased cellular caspase-3 in cultured cells, however, only MDM2, but not MDMX, inhibition results in a significant p53 protein level upregulation.
Packaging
Packaged under inert gas
Preparation Note
Following reconstitution, aliquot and freeze (-20°C. Stock solutions are stable for up to 6 months at -20°C.
Other Notes
Reed, D., et al. 2010. J. Biol. Chem.285, 10786.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
Lagerklasse
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.