Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 49,00 |
Über diesen Artikel
€ 49,00
material
borosilicate glass flask, pear-shaped , two-neck (angled side neck)
joint
center joint: ST/NS14/20, side joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F341105
capacity
5 mL, 5 mL
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| manufacturer/tradename Synthware F341105 | manufacturer/tradename Synthware F341115 | manufacturer/tradename Synthware F341125 | manufacturer/tradename Synthware F341110 |
| capacity 5 mL , 5 mL | capacity 15 mL , 15 mL | capacity 25 mL , 25 mL | capacity 10 mL , 10 mL |
| material borosilicate glass flask, two-neck (angled side neck), pear-shaped | material borosilicate glass flask, pear-shaped , two-neck (angled side neck) | material borosilicate glass flask, pear-shaped , two-neck (angled side neck) | material borosilicate glass flask, pear-shaped , two-neck (angled side neck) |
| joint side joint: ST/NS14/20, center joint: ST/NS14/20 | joint center joint: ST/NS14/20, side joint: ST/NS14/20 | joint center joint: ST/NS14/20, side joint: ST/NS14/20 | joint center joint: ST/NS14/20, side joint: ST/NS14/20 |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under Synthware™ two neck pear shaped flask
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?