Ansicht ändern
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 1.240,00 |
Über diesen Artikel
NACRES:
NB.61
UNSPSC Code:
81141502
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenproduct line
SynLectro®
General description
SynLectro® 200 mL Glass Cell with PTFE Stopper is the part of the SynLectro® Electrolysis Platform for synthetic electrochemistry.
Features and Benefits:
SynLectro® Electrolysis Platform
The SynLectro® Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
Features and Benefits:
- Designed to simplify setup, optimize reactivity, and improve safety
- Side-arm enables connection to reflux condenser or inert gas
- Chemical resistant material (PTFE)
- Protects electrical contacts from electrolytes
- Sleeve (optional) prevents seized joints
SynLectro® Electrolysis Platform
The SynLectro® Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
Legal Information
SynLectro is a registered trademark of Merck KGaA, Darmstadt, Germany
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description - | description - | description - | description - |
| product line SynLectro® | product line SynLectro® | product line SynLectro® | product line SynLectro® |
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?