MilliporeSigma
  • LPS-induced systemic inflammation reveals an immunomodulatory role for the prion protein at the blood-brain interface.
LPS-induced systemic inflammation reveals an immunomodulatory role for the prion protein at the blood-brain interface.
Journal of neuroinflammation (2017-05-24)
Ø Salvesen, M R Reiten, A Espenes, M K Bakkebø, M A Tranulis, C Ersdal
ABSTRACT
The cellular prion protein (PrP In order to explore putative roles for PrP All LPS-treated goats displayed clinical signs of sickness behavior, which were of significantly (p < 0.01) longer duration in animals without PrP Our data suggest that PrP
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, γ-irradiated, BioXtra, suitable for cell culture