MilliporeSigma
  • Bio-functionalization of monodisperse magnetic nanoparticles and their use as biomolecular labels in a magnetic tunnel junction based sensor.
Bio-functionalization of monodisperse magnetic nanoparticles and their use as biomolecular labels in a magnetic tunnel junction based sensor.
The journal of physical chemistry. B (2006-07-21)
Stephanie G Grancharov, Hao Zeng, Shouheng Sun, Shan X Wang, Stephen O'Brien, C B Murray, J R Kirtley, G A Held
ABSTRACT
Monodisperse magnetic nanoparticles (NPs) could enable the ultra-sensitive magnetic detection of biological analytes. However, rendering these particles biocompatible has remained a challenge. We report the bio-functionalization and detection of 12-nm manganese ferrite NPs. We have achieved the site-specific binding of biotin-functionalized NPs onto avidin-patterned silicon oxide substrates and DNA-functionalized NPs onto complementary DNA-patterned silicon oxide substrates. Utilizing scanning SQUID microscopy, we show that these substrate-bound NPs retain their magnetic properties. Finally, we demonstrate a novel method of detecting either protein binding or DNA hybridization at room temperature using the NPs and a magnetic tunnel-junction-based biosensor situated in orthogonal magnetic fields.
Sigma-Aldrich
Avidin–FITC from egg white, buffered aqueous solution