MilliporeSigma
  • BioTAP-XL: Cross-linking/Tandem Affinity Purification to Study DNA Targets, RNA, and Protein Components of Chromatin-Associated Complexes.
BioTAP-XL: Cross-linking/Tandem Affinity Purification to Study DNA Targets, RNA, and Protein Components of Chromatin-Associated Complexes.
Current protocols in molecular biology (2015-01-07)
Artyom A Alekseyenko, Kyle A McElroy, Hyuckjoon Kang, Barry M Zee, Peter V Kharchenko, Mitzi I Kuroda
ABSTRACT
In order to understand how chromatin complexes function in the nucleus, it is important to obtain a comprehensive picture of their protein, DNA, and RNA components, as well as their mutual interactions. This unit presents a chromatin cross-linking approach (BioTAP-XL) that utilizes a special BioTAP-tagged transgenic protein bait along with mass spectrometry to identify protein complex components, and high-throughput sequencing to identify RNA components and DNA binding sites. Full protocols are provided for Drosophila cells and for human cells in culture, along with an additional protocol for Drosophila embryos as the source material. A key element of the approach in all cases is the generation of control data from input chromatin samples.
Sigma-Aldrich
Peroxidase Anti-Peroxidase Soluble Complex antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Roche
Proteinase K, recombinant, PCR Grade, Lyophilizate from Pichia pastoris
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Rabbit IgG−Agarose, saline suspension
Millipore
Nylon Membrane Filter, 100.0 μm Pore Size, Millipore, 30 cm x 3 m, hydrophilic
Roche
RNase, DNase-free, High Concentration, from bovine pancreas