MilliporeSigma
A potentiometric biosensor for rapid on-site disease diagnostics.
Biosensors & bioelectronics (2016-01-15)
Alexey Tarasov, Darren W Gray, Meng-Yen Tsai, Niall Shields, Armelle Montrose, Niamh Creedon, Pierre Lovera, Alan O'Riordan, Mark H Mooney, Eric M Vogel
ABSTRACT
Quantitative point-of-care (POC) devices are the next generation for serological disease diagnosis. Whilst pathogen serology is typically performed by centralized laboratories using Enzyme-Linked ImmunoSorbent Assay (ELISA), faster on-site diagnosis would infer improved disease management and treatment decisions. Using the model pathogen Bovine Herpes Virus-1 (BHV-1) this study employs an extended-gate field-effect transistor (FET) for direct potentiometric serological diagnosis. BHV-1 is a major viral pathogen of Bovine Respiratory Disease (BRD), the leading cause of economic loss ($2 billion annually in the US only) to the cattle and dairy industry. To demonstrate the sensor capabilities as a diagnostic tool, BHV-1 viral protein gE was expressed and immobilized on the sensor surface to serve as a capture antigen for a BHV-1-specific antibody (anti-gE), produced in cattle in response to viral infection. The gE-coated immunosensor was shown to be highly sensitive and selective to anti-gE present in commercially available anti-BHV-1 antiserum and in real serum samples from cattle with results being in excellent agreement with Surface Plasmon Resonance (SPR) and ELISA. The FET sensor is significantly faster than ELISA (<10 min), a crucial factor for successful disease intervention. This sensor technology is versatile, amenable to multiplexing, easily integrated to POC devices, and has the potential to impact a wide range of human and animal diseases.
Sigma-Aldrich
Anti-Haptoglobin antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
Sigma-Aldrich
6-Mercaptohexanoic acid, 90%
Sigma-Aldrich
Haptoglobin from pooled human plasma, ≥95% (SDS-PAGE), lyophilized powder