Skip to Content
MilliporeSigma

H3536

Haptoglobin from pooled human plasma

≥95% (SDS-PAGE), lyophilized powder

Synonym(s):

Haptoglobin from human plasma

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.61
MDL number:
Form:
lyophilized powder
Assay:
≥95% (SDS-PAGE)
Biological source:
human plasma (pooled)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human plasma (pooled)

Quality Segment

assay

≥95% (SDS-PAGE)

form

lyophilized powder

technique(s)

activity assay: suitable

solubility

H2O: soluble 10 mg/mL

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... HP(3240)

General description

Haptoglobin is an α2-globulin found in normal plasma at a concentration of 82-236 mg/dl and accounts for 0.4-2.6% of the total plasma proteins.The monomeric form of this acute phase plasma protein is composed of four polypeptide chains: two α chains and two β chains.
Haptoglobin is an acute phase protein that scavenges hemoglobin released into the circulation. Haptoglobin, the plasma protein with highest binding affinity to hemoglobin, is mainly expressed in the liver.

Application

Haptoglobin was used to study induction of the acrosome reaction in sperm. Exposure to low temperature increases their rate of fusion with zona-free hamster oocytes.

Biochem/physiol Actions

Functions by binding to plasma hemoglobin (from hemolysis). The complex is removed from the circulation, by binding to CD163 receptors on macrophages, to salvage iron and prevent oxidative damage.

Other Notes

Occurs as 3 major phenotypes: type 1-1, type 2-1, type 2-2.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library