MilliporeSigma
  • Pull-down of 5-hydroxymethylcytosine DNA using JBP1-coated magnetic beads.
Pull-down of 5-hydroxymethylcytosine DNA using JBP1-coated magnetic beads.
Nature protocols (2012-01-28)
Adam B Robertson, John Arne Dahl, Rune Ougland, Arne Klungland
ABSTRACT
We describe a method for the efficient and selective identification of DNA containing the 5-hydroxymethylcytosine (5-hmC) modification. This protocol takes advantage of two proteins: T4 β-glucosyltransferase (β-gt), which converts 5-hmC to β-glucosyl-5-hmC (β-glu-5-hmC), and J-binding protein 1 (JBP1), which specifically recognizes and binds to β-glu-5-hmC. We describe the steps necessary to purify JBP1 and modify this protein such that it can be fixed to magnetic beads. Thereafter, we detail how to use the JBP1 magnetic beads to obtain DNA that is enriched with 5-hmC. This method is likely to produce results similar to those of other 5-hmC pull-down assays; however, all necessary components for the completion of this protocol are readily available or can be easily and rapidly synthesized using basic molecular biology techniques. This protocol can be completed in less than 2 weeks and allows the user to isolate 5-hmC-containing genomic DNA that is suitable for analysis by quantitative PCR (qPCR), sequencing, microarray and other molecular biology assays.
Sigma-Aldrich
Copper(II) chloride dihydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biology
Sigma-Aldrich
EZMix N-Z-Amine® A, Casein enzymatic hydrolysate, from bovine milk
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Albumin from porcine serum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)