MilliporeSigma
  • Levels and sources of volatile organic compounds including carbonyls in indoor air of homes of Puertollano, the most industrialized city in central Iberian Peninsula. Estimation of health risk.
Levels and sources of volatile organic compounds including carbonyls in indoor air of homes of Puertollano, the most industrialized city in central Iberian Peninsula. Estimation of health risk.
International journal of hygiene and environmental health (2015-05-31)
Florentina Villanueva, Araceli Tapia, Mariano Amo-Salas, Alberto Notario, Beatriz Cabañas, Ernesto Martínez
ABSTRACT
Twenty nine organic air pollutants including carbonyl compounds, alkanes, aromatic hydrocarbons and terpenes were measured in the indoor environment of different houses together with the corresponding outdoor measurements in Puertollano, the most industrialized city in central Iberian Peninsula. VOCs were sampled during 8 weeks using Radiello(®) passive samplers, and a questionnaire on potential VOCs sources was filled out by the occupants. The results show that formaldehyde and hexanal was the most abundant VOCs measured in indoor air, with a median concentration of 55.5 and 46.4μgm(-3), respectively followed by butanal (29.1μgm(-3)), acetone (28.4μgm(-3)) and acetaldehyde (21.4μgm(-3)). After carbonyls, n-dodecane (13.1μgm(-3)) and terpenes (α-pinene, 13.4μgm(-3) and limonene, 13.4μgm(-3)) were the compounds with higher median concentrations. The indoor/outdoor (I/O) ratios demonstrated that sources in the indoor environment are prevailing for most of the investigated VOCs especially for limonene, α-pinene, hexanal, formaldehyde, pentanal, acetaldehyde, o-xylene, n-dodecane and acetone with I/O ratio >6. Multiple linear regressions were applied to investigate the indoor VOC determinants and Spearman correlation coefficients were used to establish common sources between VOCs. Finally, the lifetime cancer risk associated to formaldehyde, acetaldehyde and benzene exposure was estimated and they varied from 7.8×10(-5) to 4.1×10(-4) for formaldehyde, from 8.6×10(-6) to 3.5×10(-5) for acetaldehyde and from 2.0×10(-6) to 1.5×10(-5) for benzene. For formaldehyde, the attributed risk in most sampled homes was two orders of magnitude higher than the one (10(-6)) proposed as acceptable by risk management bodies.
Product Description
radiello Filtration Kit, pk of 20
radiello Diffusive Bodies, white, configured for general use, pk of 20
Supelco
radiello BTEX Calibration Kit (CS2 Desorption)
Supelco
radiello Methylene Blue Calibration Standard for H2S (Hydrogen Sulfide), pkg of 100 mL
radiello Diffusive Bodies, yellow, configured for reduced sampling rates, pk of 20
radiello Diffusive Bodies, blue, configured for sampling light sensitive compounds, pk of 20
radiello BTEX/VOC Starter Kit, CS2 Desorption
radiello Empty Storage Tubes, polypropylene (with stopper), volume 12 mL, pk of 100
radiello On-Field Thermometer, Includes one standard Radiello vertical adapter (RAD122) for each thermometer, configured for standard use, pk of 3
radiello BTEX/VOC Starter Kit, Thermal Desorption
Sigma-Aldrich
Heptane, SAJ first grade, ≥98.0%
Sigma-Aldrich
1-Butanol, SAJ first grade, ≥99.0%
Sigma-Aldrich
1-Butanol, JIS special grade, ≥99.0%
Sigma-Aldrich
Styrene, SAJ first grade, ≥99.0%
Sigma-Aldrich
Octane, electronic grade, ≥99.999% metals basis, ≥99% (CP)
Sigma-Aldrich
1-Butanol, suitable for HPLC
Sigma-Aldrich
Heptane, JIS special grade, ≥99.0%
radiello 1,3-Butadiene Starter Kit, Thermal Desorption, pkg of 1 kit (2 RAD141 adsorbing cartridges and barcode labels, 1 RAD1202 yellow diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet)
Supelco
(−)-Δ9-THC-D3 solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Decane, ≥95%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Undecane, ≥99%
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Supelco
Heptane, HPLC grade, ≥99%
Sigma-Aldrich
Nonane, anhydrous, ≥99%
Sigma-Aldrich
Octane, anhydrous, ≥99%
Sigma-Aldrich
Undecane, ≥99%
Supelco
1-Butanol, suitable for HPLC, 99.8%
Sigma-Aldrich
m-Xylene, anhydrous, ≥99%