MilliporeSigma
  • Alirocumab as Add-On to Atorvastatin Versus Other Lipid Treatment Strategies: ODYSSEY OPTIONS I Randomized Trial.
Alirocumab as Add-On to Atorvastatin Versus Other Lipid Treatment Strategies: ODYSSEY OPTIONS I Randomized Trial.
The Journal of clinical endocrinology and metabolism (2015-06-02)
Harold Bays, Daniel Gaudet, Robert Weiss, Juan Lima Ruiz, Gerald F Watts, Ioanna Gouni-Berthold, Jennifer Robinson, Jian Zhao, Corinne Hanotin, Stephen Donahue
ABSTRACT
Despite current standard of care, many patients at high risk of cardiovascular disease (CVD) still have elevated low-density lipoprotein cholesterol (LDL-C) levels. Alirocumab is a fully human monoclonal antibody inhibitor of proprotein convertase subtilisin/kexin type 9. The objective of the study was to compare the LDL-C-lowering efficacy of adding alirocumab vs other common lipid-lowering strategies. Patients (n = 355) with very high CVD risk and LDL-C levels of 70 mg/dL or greater or high CVD risk and LDL-C of 100 mg/dL or greater on baseline atorvastatin 20 or 40 mg were randomized to one of the following: 1) add-on alirocumab 75 mg every 2 weeks (Q2W) sc; 2) add-on ezetimibe 10 mg/d; 3) double atorvastatin dose; or 4) for atorvastatin 40 mg regimen only, switch to rosuvastatin 40 mg. For patients not achieving protocol-defined LDL-C goals, the alirocumab dose was increased (blinded) at week 12 to 150 mg Q2W. The primary end point was percentage change in calculated LDL-C from baseline to 24 weeks (intent to treat). Among atorvastatin 20 and 40 mg regimens, respectively, add-on alirocumab reduced LDL-C levels by 44.1% and 54.0% (P < .001 vs all comparators); add-on ezetimibe, 20.5% and 22.6%; doubling of atorvastatin dose, 5.0% and 4.8%; and switching atorvastatin 40 mg to rosuvastatin 40 mg, 21.4%. Most alirocumab-treated patients (87.2% and 84.6%) achieved their LDL-C goals. Most alirocumab-treated patients (86%) maintained their 75-mg Q2W regimen. Treatment-emergent adverse events occurred in 65.4% of alirocumab patients vs 64.4% ezetimibe and 63.8% double atorvastatin/switch to rosuvastatin (data were pooled). Adding alirocumab to atorvastatin provided significantly greater LDL-C reductions vs adding ezetimibe, doubling atorvastatin dose, or switching to rosuvastatin and enabled greater LDL-C goal achievement.
Sigma-Aldrich
ADA, ≥98% (titration)