MilliporeSigma
  • Characterization of Shrimp Oil from Pandalus borealis by High Performance Liquid Chromatography and High Resolution Mass Spectrometry.
Characterization of Shrimp Oil from Pandalus borealis by High Performance Liquid Chromatography and High Resolution Mass Spectrometry.
Marine drugs (2015-06-23)
Guangling Jiao, Joseph P M Hui, Ian W Burton, Marie-Hélène Thibault, Claude Pelletier, Josée Boudreau, Nadia Tchoukanova, Balaji Subramanian, Yahia Djaoued, Stephen Ewart, Jacques Gagnon, Kathryn Vanya Ewart, Junzeng Zhang
ABSTRACT
Northern shrimp (Pandalus borealis) oil, which is rich in omega-3 fatty acids, was recovered from the cooking water of shrimp processing facilities. The oil contains significant amounts of omega-3 fatty acids in triglyceride form, along with substantial long-chain monounsaturated fatty acids (MUFAs). It also features natural isomeric forms of astaxanthin, a nutritional carotenoid, which gives the oil a brilliant red color. As part of our efforts in developing value added products from waste streams of the seafood processing industry, we present in this paper a comprehensive characterization of the triacylglycerols (TAGs) and astaxanthin esters that predominate in the shrimp oil by using HPLC-HRMS and MS/MS, as well as 13C-NMR. This approach, in combination with FAME analysis, offers direct characterization of fatty acid molecules in their intact forms, including the distribution of regioisomers in TAGs. The information is important for the standardization and quality control, as well as for differentiation of composition features of shrimp oil, which could be sold as an ingredient in health supplements and functional foods.
Sigma-Aldrich
Myristic acid, ≥95%, FCC, FG
Sigma-Aldrich
Myristic acid, natural, ≥98.5%, FG
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
cis-10-Heptadecenoic acid, ≥99%
Sigma-Aldrich
cis-Vaccenic acid, ≥97% (capillary GC), oil
Sigma-Aldrich
trans-Vaccenic acid, ≥99% (capillary GC)
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Myristic acid, Sigma Grade, ≥99%
Sigma-Aldrich
Linoleic acid, ≥98%
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Palmitoleic acid, ≥98.5% (GC), liquid
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Pentadecanoic acid, ~99% (capillary GC)
Sigma-Aldrich
Tricosanoic acid, ≥99% (capillary GC)