MilliporeSigma
  • A small, differentially regulated family of farnesyl diphosphate synthases in maize (Zea mays) provides farnesyl diphosphate for the biosynthesis of herbivore-induced sesquiterpenes.
A small, differentially regulated family of farnesyl diphosphate synthases in maize (Zea mays) provides farnesyl diphosphate for the biosynthesis of herbivore-induced sesquiterpenes.
Planta (2015-02-15)
Annett Richter, Irmgard Seidl-Adams, Tobias G Köllner, Claudia Schaff, James H Tumlinson, Jörg Degenhardt
ABSTRACT
Of the three functional FPPS identified in maize, fpps3 is induced by herbivory to produce FDP important for the formation of the volatile sesquiterpenes of plant defense. Sesquiterpenes are not only crucial for the growth and development of a plant but also for its interaction with the environment. The biosynthesis of sesquiterpenes proceeds over farnesyl diphosphate (FDP), which is either used as a substrate for protein prenylation, converted to squalene, or to volatile sesquiterpenes. To elucidate the regulation of sesquiterpene biosynthesis in maize, we identified and characterized the farnesyl diphosphate synthase (FPPS) gene family which consists of three genes. Synteny analysis indicates that fpps2 and fpps3 originate from a genome duplication in an ancient tetraploid ancestor. The three FPPSs encode active enzymes that produce predominantly FDP from the isopentenyl diphosphate and dimethylallyl diphosphate substrates. Only fpps1 and fpps3 are induced by elicitor treatment, but induced fpps1 levels are much lower and only increased to the amounts of fpps3 levels in intact leaves. Elicitor-induced fpps3 levels in leaves increase to more than 15-fold of background levels. In undamaged roots, transcript levels of fpps1 are higher than those of fpps3, but only fpps3 transcripts are induced in response to herbivory by Diabrotica virgifera virgifera. A kinetic of transcript abundance in response to herbivory in leaves provided further evidence that the regulation of fpps3 corresponds to that of tps23, a terpene synthase, that converts FDP to the volatile (E)-ß-caryophyllene. Our study indicates that the differential expression of fpps1 and fpps3 provides maize with FDP for both primary metabolism and terpene-based defenses. The expression of fpps3 seems to coincide with the herbivore-induced emission of volatile sesquiterpenes that were demonstrated to be important defense signals.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
(+)-Sodium L-ascorbate, SAJ special grade, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
MOPSO, ≥99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%