MilliporeSigma
  • Blood-brain barrier-permeable fluorone-labeled dieckols acting as neuronal ER stress signaling inhibitors.
Blood-brain barrier-permeable fluorone-labeled dieckols acting as neuronal ER stress signaling inhibitors.
Biomaterials (2015-05-23)
Jong Hwan Kwak, Zhigang Yang, Byungkwon Yoon, Yanxia He, Soojin Uhm, Hyeon-Cheol Shin, Bong Ho Lee, Yung Choon Yoo, Kyung Bok Lee, Seung-Yun Han, Jong Seung Kim
ABSTRACT
We studied the blood-brain barrier (BBB) permeability and intracellular localization of a fluorescein isothiocyanate (FITC)-labeled dieckol (1) and a rhodamine B-labeled dieckol (7), for exploring the possible therapeutic application of fluorone-labeled dieckols in neurodegenerative diseases. Both compounds (1 &7) were synthesized through a click reaction and were found to be localized in the endoplasmic reticulum (ER) of the two types of brain cell lines (SH-SY5Y and BV-2 cells) tested; they also reduced ER stress in the SH-SY5Y human neuroblastoma cells. In addition, 1 and 7 were shown to pass the BBB in rats upon intravenous administration. Altogether, our study demonstrates, for the first time, that targeted ER-stress reduction in brain cells can be achieved by introducing fluorone-dieckol conjugates into systemic circulation. Therefore, 1 and 7 provide a novel and promising ER-targeting therapeutic strategy for neurodegenerative diseases.
Sigma-Aldrich
Sodium azide, SAJ first grade, ≥97.0%
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Propargyl bromide solution, 80 wt. % in toluene, contains 0.3% magnesium oxide as stabilizer
Sigma-Aldrich
Propargyl bromide solution, 80 wt. % in xylene
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications