Skip to Content
MilliporeSigma

07-4875

Dimethyl sulfoxide

≥99.5%

Synonym(s):

DMSO

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
EC Number:
200-664-3
UNSPSC Code:
12191600
PubChem Substance ID:
Beilstein/REAXYS Number:
506008
MDL number:
Assay:
≥99.5%
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

assay

≥99.5%

form

liquid

autoignition temp.

573 °F

expl. lim.

42 %, 63 °F

availability

available only in Japan

refractive index

n20/D 1.479 (lit.)

bp

189 °C (lit.)

mp

16-19 °C (lit.)

density

1.10 g/mL (lit.)

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Application

for biological purposes

Disclaimer

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.


Still not finding the right product?

Explore all of our products under Dimethyl sulfoxide


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

188.6 °F - closed cup

flash_point_c

87 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library