MilliporeSigma
  • Simultaneous determination of bambuterol and its two major metabolites in human plasma by hydrophilic interaction ultra-performance liquid chromatography-tandem mass spectrometry.
Simultaneous determination of bambuterol and its two major metabolites in human plasma by hydrophilic interaction ultra-performance liquid chromatography-tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-08-17)
Ting Zhou, Qing Cheng, Chengjuan Zou, Ting Zhao, Shan Liu, Marco Pistolozzi, Evina Tan, Ling Xu, Wen Tan
ABSTRACT
In this study, a rapid and sensitive hydrophilic interaction ultra-performance liquid chromatography-tandem mass spectrometry (HILIC-UPLC-MS/MS) method was developed for simultaneous determination of bambuterol and its two major metabolites monocarbamate bambuterol and terbutaline in human plasma. All samples were simply precipitated using acetonitrile and separated on a UPLC-HILIC column under gradient elution with a mobile phase consisting of acetonitrile and water with the addition of 10mm ammonium acetate and 0.1% formic acid at 0.4 mL/min. The analytes were detected by a Xevo TQ-S tandem mass spectrometer with positive electrospray ionization in multiple reaction monitoring mode. The established method was highly sensitive with the lower limit of quantification (LLOQ) of 10.00 pg/mL for each analyte, and the intra- and inter-day precisions were <12.8%. The analytical runtime within 4.0 min per sample made this method suitable for high throughput determination. The validated method was successfully applied to a clinical pharmacokinetic study of bambuterol in eight healthy volunteers. Furthermore, the effects of the chromatographic conditions on the retention of the analytes on HILIC were investigated, and the benefits of HILIC were evaluated by comparing with a C18 column. The results indicated that liquid-liquid partition and the electrostatic interactions played an important role in the retention of the analytes on HILIC in this study. And HILIC offered particular advantages over RPLC approach in the aspects of the peak symmetry, the column efficiency, and the column pressure.
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Sigma-Aldrich
Ethyl acetate, suitable for HPLC
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard