MilliporeSigma
  • Slug is temporally regulated by cyclin E in cell cycle and controls genome stability.
Slug is temporally regulated by cyclin E in cell cycle and controls genome stability.
Oncogene (2014-03-26)
W-L Wang, H-C Huang, S-H Kao, Y-C Hsu, Y-T Wang, K-C Li, Y-J Chen, S-L Yu, S-P Wang, T-H Hsiao, P-C Yang, T-M Hong
ABSTRACT
The transcriptional repressor Slug is best known to control epithelial-mesenchymal transition (EMT) and promote cancer invasion/metastasis. In this study, we demonstrate that Slug is temporally regulated during cell cycle progression. At G1/S transition, cyclin E-cyclin-dependent kinase 2 mediates the phosphorylation of Slug at Ser-54 and Ser-104, resulting in its ubiquitylation and degradation. Non-phosphorylatable Slug is markedly stabilized at G1/S transition compared with wild-type Slug and greatly leads to downregulation of DNA synthesis and checkpoint-related proteins, including TOP1, DNA Ligase IV and Rad17, reduces cell proliferation, delays S-phase progression and contributes to genome instability. Our results indicate that Slug has multifaceted roles in cancer progression by controlling both EMT and genome stability.
Product Description
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis