MilliporeSigma
  • Metabolomics study of hepatocellular carcinoma: discovery and validation of serum potential biomarkers by using capillary electrophoresis-mass spectrometry.
Metabolomics study of hepatocellular carcinoma: discovery and validation of serum potential biomarkers by using capillary electrophoresis-mass spectrometry.
Journal of proteome research (2014-05-24)
Jun Zeng, Peiyuan Yin, Yexiong Tan, Liwei Dong, Chunxiu Hu, Qiang Huang, Xin Lu, Hongyang Wang, Guowang Xu
ABSTRACT
Hepatocellular carcinoma (HCC) is one of the most lethal malignancies. The lack of effective screening methods for early diagnosis has been a longstanding bottleneck to improve the survival rate. In the present study, a capillary electrophoresis-time-of-flight mass spectrometry (CE-TOF/MS)-based metabolomics method was employed to discover novel biomarkers for HCC. A total of 183 human serum specimens (77 sera in discovery set and 106 sera in external validation set) were enrolled in this study, and a "serum biomarker model" including tryptophan, glutamine, and 2-hydroxybutyric acid was finally established based on the comprehensive screening and validation workflow. This model was evaluated as an effective tool in that area under the receiver operating characteristic curve reached 0.969 in the discovery set and 0.99 in the validation set for diagnosing HCC from non-HCC (health and cirrhosis). Furthermore, this model enabled the discrimination of small HCC from precancer cirrhosis with an AUC of 0.976, highlighting the potential of early diagnosis. The biomarker model is effective for those a-fetoprotein (AFP) false-negative and false-postive subjects, indicating the complementary function to conventional tumor marker AFP. This study demonstrates the promising potential of CE-MS-based metabolomics approach in finding biomarkers for disease diagnosis and providing special insights into tumor metabolism.
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
Succinic acid-13C4, 99 atom % 13C
Sigma-Aldrich
L-Alanine-3,3,3-d3, 99 atom % D
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
L-Methionine sulfone
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard