MilliporeSigma
  • Endogenous functional compounds in Korean native chicken meat are dependent on sex, thermal processing and meat cut.
Endogenous functional compounds in Korean native chicken meat are dependent on sex, thermal processing and meat cut.
Journal of the science of food and agriculture (2014-08-27)
Dinesh D Jayasena, Samooel Jung, Sun Hyo Kim, Hyun Joo Kim, Amali U Alahakoon, Jun Heon Lee, Cheorun Jo
ABSTRACT
In this study the effects of sex, meat cut and thermal processing on the carnosine, anserine, creatine, betaine and carnitine contents of Korean native chicken (KNC) meat were determined. Forty 1-day-old chicks (20 chicks of each sex) from a commercial KNC strain (Woorimatdag™) were reared under similar standard commercial conditions with similar diets, and ten birds of each sex were randomly selected and slaughtered at 14 weeks of age. Raw and cooked meat samples were prepared from both breast and leg meats and analyzed for the aforementioned functional compounds. Female KNCs had significantly higher betaine and creatine contents. The breast meat showed significantly higher carnosine and anserine contents, whereas the leg meat had a higher betaine and carnitine content. The content of all functional compounds was significantly depleted by thermal processing. This study confirms that KNC meat is a good source of the above-mentioned functional compounds, which can be considered attractive nutritional quality factors. However, their concentrations were significantly affected by thermal processing conditions, meat cut and sex. Further experiments are needed to select the best thermal processing method to preserve these functional compounds.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Creatine, anhydrous
Sigma-Aldrich
(±)-Carnitine hydrochloride, ≥98% (TLC)
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
L-Carnosine, ~99%, crystalline
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol