MilliporeSigma
  • Pharmacokinetics of dexamethasone and nefopam administered alone or in combination using a newly developed prefilled multi-drug injector in rats.
Pharmacokinetics of dexamethasone and nefopam administered alone or in combination using a newly developed prefilled multi-drug injector in rats.
Pharmacology (2014-07-12)
Jie Ma, Junhai Zhang, Tao Yang, Kaihua Fan, Jianwen Gu, Guangfu Yin
ABSTRACT
It is significant for patients with traumatic brain injury (TBI) to receive prehospital emergency treatment as early as possible to reduce mortality. Therefore, a new prefilled multi-drug injector was developed to improve the treatment of TBI. Here, we studied the pharmacokinetics of dexamethasone (DXM) and nefopam using the injector to investigate the significance of drug interactions and the necessity of dose adjustment. Rats were administered DXM and nefopam intramuscularly alone or in combination using the injector. The concentrations of DXM and nefopam were measured by means of HPLC. The noncompartmental approach was used to calculate pharmacokinetic parameters. All animals appeared to tolerate the injection very well. The maximum concentration 90% confidence interval (CI) of nefopam was in the bioequivalence range when nefopam was co-administered with DXM. However, the AUC 90% CI of nefopam was out of the range. A statistically significant alteration was also observed in the clearance of nefopam. The co-administration exhibited no significant influence on the pharmacokinetic parameters of DXM. These results indicate that the co-administration of DXM and nefopam using the prefilled multi-drug injector significantly alters some pharmacokinetic parameters of nefopam and has a minor effect on DXM pharmacokinetics.
Sigma-Aldrich
Sodium 1-heptanesulfonate, BioXtra
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Sodium 1-heptanesulfonate
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium 1-heptanesulfonate, Redi-Dri, free-flowing
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C