MilliporeSigma
  • Isolation, chemical characterization, and free radical scavenging activity of phenolics from Triticum aestivum L. aerial parts.
Isolation, chemical characterization, and free radical scavenging activity of phenolics from Triticum aestivum L. aerial parts.
Journal of agricultural and food chemistry (2014-10-31)
Iwona Kowalska, Lukasz Pecio, Lukasz Ciesla, Wieslaw Oleszek, Anna Stochmal
ABSTRACT
Fourteen phenolic compounds (flavonoids and phenolic acids) were isolated and 19 were identified in the aerial parts of Triticum aestivum L. The structures of these compounds were established on the basis of the data obtained by electrospray ionization-mass spectrometry (ESI-MS) and nuclear magnetic resonance (NMR) techniques. T. aestivum L. was found to be rich in flavones, especially in luteolin derivatives. Three of the isolated compounds, including luteolin 6-C-[6Glc″-O-E-caffeoyl-β-D-glucopyranosyl(1″→2)-β-glucopyranoside], luteolin 6-C-[5Rib″-O-E-feruoyl-β-D-ribofuranosyl(1″→2)-β-glucopyranoside], and 3',4',5'-O-trimethyltricetin 7-O-[β-D-glucuropyranosyl(1″→2)-β-D-glucopyranoside], have been reported for the first time in the plant kingdom. The amount of individual phenolics, in winter wheat, was also determined. Additionally, the free radical scavenging potential of the isolated compounds was tested in a simple and rapid thin-layer chromatography-2,2-diphenyl-1-picrylhydrazyl radical test (TLC-DPPH•) with image processing.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Chloroform, suitable for HPLC
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%