MilliporeSigma
  • The acylated (AG) to unacylated (UAG) ghrelin ratio in esterase inhibitor-treated blood is higher than previously described.
The acylated (AG) to unacylated (UAG) ghrelin ratio in esterase inhibitor-treated blood is higher than previously described.
Clinical endocrinology (2014-05-09)
Patric J D Delhanty, Martin Huisman, Michel Julien, Karine Mouchain, Patrick Brune, Axel P N Themmen, Thierry Abribat, Aart J van der Lely
ABSTRACT
The acylated/unacylated ghrelin (AG/UAG) ratio has been reported to range from 0·02 to 0·3, suggesting biologically relevant independent regulation of each ghrelin isoform. However, AG is deacylated to UAG by esterases in blood samples, and esterase inhibition is critical for their accurate measurement. Our hypothesis is that at least part of the variation in reported AG and UAG values is due to inconsistent sample preparation. A non-interventional study. Quantification with two different, commercially available, ELISA formats of AG and UAG in venous plasma stabilized or not with 4-(2-aminoethyl) benzenesulphonyl fluoride (AEBSF) and stored for 0-6 months at -20 or -80 °C. Healthy, non-obese, adults (n = 8; 4 women), age 26-42 yrs, after an overnight fast. AG and UAG stability following different methods of sample treatment and storage. Non-AEBSF plasma contained low AG and high UAG (>270 pg/ml) indicating rapid conversion of AG to UAG. However, AEBSF plasma, stored at -80 °C and measured at 0, 1, 3 and 6 months contained AG and UAG ranges of 12-350 and 17-170 pg/ml, respectively. Mean (SEM) AG/UAG ratios were 1·7(0·3), 1·2(0·2), 1·5(0·3) and 1·8(0·5) at each time point with no significant effect of storage period. AG and UAG levels measured in AEBSF-stabilized plasma indicate that the AG/UAG ratio is markedly higher than previously described and that UAG is a physiological component of the circulation. This highlights the importance of immediately stabilizing blood samples on collection for determination of both AG and UAG concentrations and provides a valuable tool for their measurement in physiological and interventional studies.
Product Description
Supelco
Pefabloc® SC, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Roche
Pefabloc® SC, powder, solubility: 100 mg/mL in aqueous buffer, suitable for blocking, suitable for protein purification