MilliporeSigma
  • Atmospheric pressure gas chromatography-time-of-flight-mass spectrometry (APGC-ToF-MS) for the determination of regulated and emerging contaminants in aqueous samples after stir bar sorptive extraction (SBSE).
Atmospheric pressure gas chromatography-time-of-flight-mass spectrometry (APGC-ToF-MS) for the determination of regulated and emerging contaminants in aqueous samples after stir bar sorptive extraction (SBSE).
Analytica chimica acta (2014-12-03)
Marina G Pintado-Herrera, Eduardo González-Mazo, Pablo A Lara-Martín
ABSTRACT
This work presents the development, optimization and validation of a multi-residue method for the simultaneous determination of 102 contaminants, including fragrances, UV filters, repellents, endocrine disruptors, biocides, polycyclic aromatic hydrocarbons (PAHs), polychlorinated biphenyls (PCBs), and several types of pesticides in aqueous matrices. Water samples were processed using stir bar sorptive extraction (SBSE) after the optimization of several parameters: agitation time, ionic strength, presence of organic modifiers, pH, and volume of the derivatizing agent. Target compounds were extracted from the bars by liquid desorption (LD). Separation, identification and quantification of analytes were carried out by gas chromatography (GC) coupled to time-of-flight (ToF-MS) mass spectrometry. A new ionization source, atmospheric pressure gas chromatography (APGC), was tested. The optimized protocol showed acceptable recovery percentages (50-100%) and limits of detection below 1ngL(-1) for most of the compounds. Occurrence of 21 out of 102 analytes was confirmed in several environmental aquatic matrices, including seawater, sewage effluent, river water and groundwater. Non-target compounds such as organophosphorus flame retardants were also identified in real samples by accurate mass measurement of their molecular ions using GC-APGC-ToF-MS. To the best of our knowledge, this is the first time that this technique has been applied for the analysis of contaminants in aquatic systems. By employing lower energy than the more widely used electron impact ionization (EI), AGPC provides significant advantages over EI for those substances very susceptible to high fragmentation (e.g., fragrances, pyrethroids).
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetic anhydride, JIS special grade
Sigma-Aldrich
Ethyl acetate, suitable for HPLC
Supelco
Octisalate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Octocrylene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cypermethrin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Supelco
Baythroid®, PESTANAL®, analytical standard
Sigma-Aldrich
Dichloromethane, JIS special grade, ≥99.0%
Sigma-Aldrich
Dichloromethane, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Ethyl acetate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethyl acetate, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Deltamethrin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Bisphenol A, ≥99%
Sigma-Aldrich
Bisphenol A, 97%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
3-(4-Methylbenzylidene)camphor, ≥98.0% (GC)
Sigma-Aldrich
Fenvalerate, ≥97%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
2-Hydroxy-4-methoxybenzophenone, 98%
Supelco
2-Ethylhexyl salicylate, analytical standard
Supelco
Octocrylene, analytical standard
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol